Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2700038G22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045040.1 | 4 | 974 | |||
NR_045042.1 | 4 | 955 | ||||
GenBank mRNA | AA163134.1 | - | 1 | 411 | ||
AK002390.1 | - | 4 | 975 | |||
AK012332.1 | - | 1 | 851 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DT916085.1 | - | 1 | 411 | Chr. 5 - 23854235 - 23854772 [+] on Build GRCm38 |