Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020445.1 | - | 2 | 1061 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA872730.1 | - | 2 | 345 | Chr. 4 - 41454391 - 41460094 [-] on Build GRCm38 |