Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013969.1 | - | 1 | 1383 | ||
AK013971.1 | - | 1 | 1387 | |||
AK170049.1 | - | 1 | 1575 |