Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052407.1 | - | 1352 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB481775.2 | - | 1 | 437 | Chr. 6 - 29761894 - 29762508 [+] on Build GRCm38 |