Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700061E18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006851.1 | - | 2 | 424 | ||
AV204968.1 | - | 1 | 124 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706834.1 | - | 2 | 425 | Chr. 13 - 41800783 - 41801791 [+] on Build GRCm38 |