Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006088.1 | - | 2 | 800 | ||
BC055348.1 | - | 2 | 595 | |||
BC059711.1 | - | 2 | 460 |