Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc75aos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019771.1 | - | 1 | 128 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715763.1 | - | 1 | 128 | Chr. 11 - 62634702 - 62638734 [+] on Build GRCm38 |