Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4833447P13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019531.1 | - | 2244 | |||
AK042445.1 | - | 1857 | ||||
BB242542.2 | - | 177 |