Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4833447P13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019531.1 | - | 793 | |||
AK034771.1 | - | 1 | 798 | |||
AK042445.1 | - | 675 |