Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm41609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082862.1 | - | 1367 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK033258.1 | - | 1429 | Chr. - - [-] on Build |