Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm2447 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087699.1 | - | 64 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB608908.2 | - | 1 | 64 | Chr. 3 - 52764600 - 52775274 [+] on Build GRCm38 |