Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D930030D11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK053103.1 | - | 471 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB663646.1 | - | 471 | Chr. - - [-] on Build |