Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810039B14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012887.1 | - | 569 | |||
AK084954.1 | - | 567 | ||||
AK171817.1 | - | 214 |