Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnmt3bos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033055.1 | - | 1 | 136 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB624052.1 | - | 1 | 129 | Chr. 2 - 153648803 - 153650280 [-] on Build GRCm38 |