Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcas3os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045875.1 | 1 | 30 | |||
GenBank mRNA | AK018376.1 | - | 1 | 31 | ||
AK053124.1 | - | 1 | 31 |
Gene Symbol: Gm10372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137618.1 | - | 332 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720493.1 | - | 1 | 31 | Chr. 11 - 85704289 - 85719744 [-] on Build GRCm38 |