Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BE691133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132899.1 | - | 1 | 851 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV265157.2 | - | 1 | 284 | Chr. 4 - 137992121 - 137992721 [-] on Build GRCm38 |