Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017601.1 | - | 1 | 88 | ||
AK021120.1 | - | 1 | 267 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY728248.1 | - | 1 | 265 | Chr. 18 - 69592604 - 69593075 [+] on Build GRCm38 |