Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700110K17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040728.1 | 2 | 430 | |||
GenBank mRNA | AK007162.1 | - | 2 | 433 | ||
BB617532.1 | - | 411 |