Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020223.1 | - | 1 | 361 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720465.1 | - | 1 | 361 | Chr. 1 - 86857371 - 86858005 [+] on Build GRCm38 |