Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102631757 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110502.1 | 4, 5 | 620 | |||
GenBank mRNA | AK085535.1 | - | 4, 5 | 621 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB502343.2 | - | 3, 4 | 341 | Chr. 17 - 31153928 - 31162323 [+] on Build GRCm38 |