Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028076.1 | - | 2 | 114 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706277.1 | - | 2 | 114 | Chr. 11 - 76217079 - 76217515 [-] on Build GRCm38 |