Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933411D12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016766.1 | - | 1 | 888 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DV043294.1 | - | 1 | 698 | Chr. 8 - 13120579 - 13121360 [+] on Build GRCm38 |