Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102640562 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK084494.1 | - | 7 | 958 |
Gene Symbol: BB218582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BB218582.1 | - | 2 | 244 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB474543.2 | - | 4 | 596 | Chr. 2 - 106642921 - 106646479 [-] on Build GRCm38 |