Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E430022K19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040137.1 | - | 1 | 1387 | ||
AK088651.1 | - | 2 | 3152 |
Gene Symbol: Glcci1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029524.1 | - | 1 | 29 | ||
BC050802.1 | - | 1 | 65 | 6771009 |