Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029096.1 | - | 2220 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB009722.2 | - | 1 | 507 | Chr. 16 - 18580980 - 18581615 [-] on Build GRCm38 |