Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046081.1 | 3 | 479 | |||
GenBank mRNA | AK144886.1 | - | 3 | 479 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY734560.1 | - | 3 | 479 | Chr. 12 - 86169341 - 86192644 [+] on Build GRCm38 |