Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011205.1 | - | 1 | 139 | ||
AK143579.1 | - | 1 | 1032 | |||
AK149041.1 | - | 1 | 624 | |||
AK169352.1 | - | 1 | 1038 |