Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2900008C10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045434.1 | 3 | 325 | |||
GenBank mRNA | BC027704.1 | - | 3 | 325 |