Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm16938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045969.1 | 4 | 981 | |||
NR_045970.1 | 4 | 957 | ||||
NR_045971.1 | 4 | 954 | ||||
NR_045972.1 | 3 | 845 | ||||
GenBank mRNA | AK085722.1 | - | 846 | |||
BC030866.1 | - | 1 | 83 | |||
BC040816.1 | - | 1 | 71 | |||
BC144932.1 | - | 4 | 934 | |||
BC144933.1 | - | 4 | 907 | |||
BC144936.1 | - | 4 | 910 |