Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK078431.1 | - | 1520 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB053280.2 | - | 1 | 507 | Chr. 3 - 122016046 - 122016751 [+] on Build GRCm38 |