Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mannr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110437.1 | 1 | 112 | |||
GenBank mRNA | AK147070.1 | - | 1 | 116 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY740722.1 | - | 1 | 116 | Chr. 3 - 29891014 - 29923401 [+] on Build GRCm38 |