Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020822.1 | - | 1 | 675 | ||
AK144564.1 | - | 1 | 1130 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB183663.2 | - | 1 | 569 | Chr. 16 - 36072186 - 36073114 [+] on Build GRCm38 |