Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI847908.1 | - | 1 | 247 | ||
AK038924.1 | - | 792 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB178800.2 | - | 1 | 244 | Chr. 4 - 19607796 - 19608409 [+] on Build GRCm38 |