Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018643.1 | - | 493 | |||
AK041061.1 | - | 1156 | ||||
AK078461.1 | - | 865 | ||||
AK165215.1 | - | 254 |