Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4833445I07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076537.1 | - | 1 | 1427 | ||
AV253134.2 | - | 1 | 437 |
Gene Symbol: Dalrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK217499.1 | - | 1 | 248 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA275329.1 | - | 1 | 74 | Chr. 9 - 108568886 - 108569174 [+] on Build GRCm38 |