Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vipas39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020129.1 | - | 1 | 521 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB679645.1 | - | 11 | Chr. 12 - 87265400 - 87265817 [-] on Build GRCm38 |