Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110018F16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003775.1 | - | 1 | 594 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB404142.2 | - | 1 | 121 | Chr. 3 - 137915899 - 137916554 [+] on Build GRCm38 |