Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam110a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080248.1 | - | 208 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB637668.1 | - | 2 | 205 | Chr. 2 - 151866231 - 151980174 [-] on Build GRCm38 |