Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sstr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK051189.1 | - | 1816 | |||
AK158660.1 | - | 483 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ144546.1 | - | 1 | 483 | Chr. 2 - 148399847 - 148400399 [+] on Build GRCm38 |