Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm7596 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK145172.1 | - | 30 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY735114.1 | - | 1 | 27 | Chr. 5 - 32390173 - 32393303 [-] on Build GRCm38 |