Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930441H08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019606.1 | - | 1, 2 | 261 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714906.1 | - | 1, 2 | 258 | Chr. 7 - 71909697 - 71939191 [+] on Build GRCm38 |