Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700006J14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005678.1 | - | 1 | 27 | ||
AK011105.1 | - | 1 | 19 | |||
BC098228.1 | - | 1 | 25 |
Gene Symbol: Hmga2-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037996.1 | 1 | 16 | |||
GenBank mRNA | AK033703.1 | - | 1 | 17 |