Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700006J14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011105.1 | - | 3, 4 | 335 | ||
BC098228.1 | - | 3, 4 | 341 |