Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_036631.2 | 6 | 1957 | |||
GenBank mRNA | AF365932.1 | - | LIT1822 Zim3 |
6 | 1962 |