Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm16206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035416.1 | - | 1890 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB112608.2 | - | 2 | 600 | Chr. 3 - 53453908 - 53457793 [+] on Build GRCm38 |