Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310034G01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040418.1 | 1 | 367 | |||
GenBank mRNA | AK009611.1 | - | 1 | 334 | ||
AK051784.1 | - | 1 | 354 |