Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700029B22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040531.1 | 5 | 665 | |||
GenBank mRNA | AK006482.1 | - | 5 | 661 | ||
BC048662.1 | - | 5 | 652 |