Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700029B22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040531.1 | 5 | 623 | |||
GenBank mRNA | AK006482.1 | - | 5 | 619 | ||
BC048662.1 | - | 5 | 610 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU937841.1 | - | 5 | 613 | Chr. 7 - 131146437 - 131150572 [+] on Build GRCm38 |