Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia3os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045366.1 | 5 | 499 | |||
GenBank mRNA | BC048666.1 | - | 5 | 499 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA465726.1 | - | 5 | 502 | Chr. 18 - 64157739 - 64277180 [-] on Build GRCm38 |