Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310001K20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009069.1 | - | 1 | 698 |
Gene Symbol: Gm40399 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK138386.1 | - | 2 | 568 |
Gene Symbol: Gm42245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029039.1 | - | 1 | 3177 |